siRNA Details
| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012767.2 | NP_036899.1 | 4 | 424 | ||
| XM_006252096.2 | XP_006252158.1 | 3 | 425 | |||
| GenBank mRNA | M12579.1 | - | 4 | 424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnrh1| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012767.2 | NP_036899.1 | 4 | 424 | ||
| XM_006252096.2 | XP_006252158.1 | 3 | 425 | |||
| GenBank mRNA | M12579.1 | - | 4 | 424 | ||