siRNA Details
| Gene Symbol: Ceacam6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB434131.1 | - | 2 | 354 | ||
| BC078962.1 | - | 2 | 593 | 7111028 | ||
| Gene Symbol: LOC103689938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759078.1 | XP_008757300.1 | 636 | |||

Ceacam6
Genomic Map