siRNA Details
| Gene Symbol: Nceh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127524.2 | NP_001120996.1 | 2 | 278 | ||
| GenBank mRNA | BC166533.1 | - | 2 | 278 | 7190480 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nceh1| Gene Symbol: Nceh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127524.2 | NP_001120996.1 | 2 | 278 | ||
| GenBank mRNA | BC166533.1 | - | 2 | 278 | 7190480 | |