siRNA Details
| Gene Symbol: Adamtsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108071.1 | NP_001101541.1 | 6 | 583 | ||
| XM_006240915.2 | XP_006240977.1 | 8 | 1512 | |||
| XM_006240916.2 | XP_006240978.1 | 8 | 1502 | |||
| XM_006240917.2 | XP_006240979.1 | 8 | 1509 | |||
| XM_006240918.2 | XP_006240980.1 | 8 | 1452 | |||
| XM_006240919.2 | XP_006240981.1 | 8 | 1511 | |||
| XM_006240920.2 | XP_006240982.1 | 7 | 1341 | |||
| GenBank mRNA | FQ224249.1 | - | 8 | 812 | ||

Adamtsl5
Genomic Map