siRNA Details
| Gene Symbol: Cer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115031.1 | NP_001108503.1 | 1 | 361 | ||
| GenBank mRNA | BC158726.1 | - | 1 | 361 | 9091451 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cer1| Gene Symbol: Cer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115031.1 | NP_001108503.1 | 1 | 361 | ||
| GenBank mRNA | BC158726.1 | - | 1 | 361 | 9091451 | |