siRNA Details
Gene Symbol: Spink4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008871.1 | NP_001008871.1 | 2 | 148 | ||
XM_006238095.2 | XP_006238157.1 | 3 | 1046 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spink4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008871.1 | NP_001008871.1 | 2 | 148 | ||
XM_006238095.2 | XP_006238157.1 | 3 | 1046 |