siRNA Details
| Gene Symbol: Asun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109608.1 | NP_001103078.1 | 14, 15 | 2284 | ||
| XM_006237689.1 | XP_006237751.1 | 14, 15 | 2329 | |||
| GenBank mRNA | FQ221916.1 | - | 4, 5 | 643 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asun| Gene Symbol: Asun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109608.1 | NP_001103078.1 | 14, 15 | 2284 | ||
| XM_006237689.1 | XP_006237751.1 | 14, 15 | 2329 | |||
| GenBank mRNA | FQ221916.1 | - | 4, 5 | 643 | ||