siRNA Details
| Gene Symbol: HAUS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015302.1 | NP_056117.1 | 19 | 2291 | ||
| XM_011526684.2 | XP_011524986.1 | 18 | 2178 | |||
| GenBank mRNA | AB020648.1 | - | 19 | 2264 | ||
| BC013947.2 | - | 19 | 2263 | |||
| BC064390.1 | - | 17 | 2480 | |||
| BC157882.1 | - | 19 | 2282 | 8862704 | ||
| BC171905.1 | - | 18 | 2536 | |||

HAUS5
Genomic Map