siRNA Details
| Gene Symbol: Tomm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135174.1 | NP_001128646.1 | 3 | 297 | ||
| GenBank mRNA | CB693792.1 | - | 3 | 307 | ||
| CB698700.1 | - | 3 | 274 | |||
| FQ221229.1 | - | 3 | 302 | |||
| FQ223031.1 | - | 3 | 303 | |||
| FQ223270.1 | - | 3 | 309 | |||
| FQ224596.1 | - | 3 | 301 | |||

Tomm7
Genomic Map