siRNA Details
| Gene Symbol: Tomm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135174.1 | NP_001128646.1 | 3 | 337 | ||
| GenBank mRNA | CB698700.1 | - | 3 | 314 | ||
| FQ221229.1 | - | 3 | 342 | |||
| FQ223031.1 | - | 3 | 343 | |||
| FQ223270.1 | - | 3 | 349 | |||
| FQ224596.1 | - | 3 | 341 | |||

Tomm7
Genomic Map