siRNA Details
| Gene Symbol: C1ql3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109403.2 | NP_001102873.1 | 2 | 680 | ||
| XM_008771889.1 | XP_008770111.1 | 4 | 5139 | |||
| XM_008771890.1 | XP_008770112.1 | 5 | 8197 | |||
| XM_008771891.1 | XP_008770113.1 | 4 | 4462 | |||

C1ql3
Genomic Map