siRNA Details
| Gene Symbol: Zfp867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109677.2 | NP_001103147.2 | 4 | 451 | ||
| GenBank mRNA | BC129129.1 | - | 4 | 451 | 7366308 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp867| Gene Symbol: Zfp867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109677.2 | NP_001103147.2 | 4 | 451 | ||
| GenBank mRNA | BC129129.1 | - | 4 | 451 | 7366308 | |