siRNA Details
| Gene Symbol: VPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009921.2 | NP_001009921.1 | 14 | 1257 | ||
| NM_015303.3 | NP_056118.2 | 13 | 1251 | |||
| XM_005247251.3 | XP_005247308.1 | 14 | 1525 | |||
| XM_005247253.4 | XP_005247310.1 | 14 | 1263 | |||
| XM_006713556.3 | XP_006713619.1 | 6 | 545 | |||
| XM_011512600.2 | XP_011510902.1 | 6 | 459 | |||
| XM_011512601.2 | XP_011510903.1 | 6 | 545 | |||
| XM_017006040.1 | XP_016861529.1 | 12 | 945 | |||
| XM_017006041.1 | XP_016861530.1 | 14 | 1263 | |||
| GenBank mRNA | AB018347.1 | - | 6 | 436 | ||
| AK292423.1 | - | 13 | 1223 | |||
| BC140768.1 | - | 14 | 1193 | 9021636 | ||

VPS8
Genomic Map