siRNA Details
| Gene Symbol: Ubxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107086.1 | NP_001100556.1 | 3 | 295 | ||
| Gene Symbol: LOC100911374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751074.3 | XP_003751122.2 | 1 | 444 | ||
| XM_008768806.1 | XP_008767028.1 | 1 | 443 | |||

Ubxn7
Genomic Map