siRNA Details
| Gene Symbol: Fsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106918.1 | NP_001100388.1 | 3, 4 | 409 | ||
| GenBank mRNA | BC160924.1 | - | 249 | 8362685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fsd1| Gene Symbol: Fsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106918.1 | NP_001100388.1 | 3, 4 | 409 | ||
| GenBank mRNA | BC160924.1 | - | 249 | 8362685 | ||