siRNA Details
| Gene Symbol: Prss33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109027.1 | NP_001102497.1 | 4 | 317 | ||
| XM_006245874.2 | XP_006245936.1 | 5 | 607 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss33| Gene Symbol: Prss33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109027.1 | NP_001102497.1 | 4 | 317 | ||
| XM_006245874.2 | XP_006245936.1 | 5 | 607 | |||