siRNA Details
Gene Symbol: Fam185a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128188.1 | NP_001121660.1 | 2, 3 | 779 | ||
GenBank mRNA | BC161871.1 | - | 2, 3 | 779 | 7112030 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam185a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128188.1 | NP_001121660.1 | 2, 3 | 779 | ||
GenBank mRNA | BC161871.1 | - | 2, 3 | 779 | 7112030 |