siRNA Details
| Gene Symbol: Olr1520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000714.1 | NP_001000714.1 | 1 | 522 | ||
| Gene Symbol: Olr1519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000037.1 | NP_001000037.1 | 1 | 522 | ||
| XM_008767928.1 | XP_008766150.1 | 1 | 462 | |||

Olr1520
Genomic Map