siRNA Details
| Gene Symbol: FAM189A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015307.1 | NP_056122.1 | 11 | 1441 | ||
| XM_011521407.2 | XP_011519709.1 | 12 | 1598 | |||
| GenBank mRNA | AB011146.1 | - | 8 | 1041 | ||
| AK295622.1 | - | 3 | 435 | |||
| DA327746.1 | - | 3 | 439 | |||

FAM189A1
Genomic Map