siRNA Details
| Gene Symbol: Tmie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109299.1 | NP_001102769.1 | 5 | 1598 | ||
| XM_006243995.2 | XP_006244057.1 | 3 | 1732 | |||
| XM_006243996.2 | XP_006244058.1 | 3 | 1374 | |||
| XM_008766649.1 | XP_008764871.1 | 3 | 1764 | |||

Tmie
Genomic Map