siRNA Details
| Gene Symbol: Olr1750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212493.1 | NP_997658.1 | 1 | 593 | ||
| Gene Symbol: Olr1746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001427.1 | NP_001001427.1 | 1 | 593 | ||

Olr1750
Genomic Map