siRNA Details
| Gene Symbol: Ryr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032078.2 | NP_114467.1 | 84 | 11647 | ||
| GenBank mRNA | EU346200.2 | - | 84 | 11647 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ryr2| Gene Symbol: Ryr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032078.2 | NP_114467.1 | 84 | 11647 | ||
| GenBank mRNA | EU346200.2 | - | 84 | 11647 | ||