siRNA Details
Gene Symbol: Ttll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108640.2 | NP_001102110.2 | 5 | 498 | ||
GenBank mRNA | BC168907.1 | - | 4 | 356 | ||
CV126544.1 | - | 5 | 608 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108640.2 | NP_001102110.2 | 5 | 498 | ||
GenBank mRNA | BC168907.1 | - | 4 | 356 | ||
CV126544.1 | - | 5 | 608 |