siRNA Details
| Gene Symbol: Tp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234865.2 | XP_006234927.1 | 25 | 5946 | ||
| XM_006234867.2 | XP_006234929.1 | 23 | 5338 | |||
| XM_008762206.1 | XP_008760428.1 | 24 | 5458 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tp53bp1| Gene Symbol: Tp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234865.2 | XP_006234927.1 | 25 | 5946 | ||
| XM_006234867.2 | XP_006234929.1 | 23 | 5338 | |||
| XM_008762206.1 | XP_008760428.1 | 24 | 5458 | |||