siRNA Details
| Gene Symbol: Tp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234865.2 | XP_006234927.1 | 23, 24 | 5542 | ||
| XM_006234867.2 | XP_006234929.1 | 21, 22 | 4934 | |||
| XM_008762206.1 | XP_008760428.1 | 22, 23 | 5054 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013735, AJ414734, AK132323, AK036324, BC035206, and others.

Tp53bp1
Genomic Map