siRNA Details
| Gene Symbol: Calu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033898.1 | NP_001029070.1 | 5 | 639 | ||
| NM_022535.2 | NP_071980.2 | 5 | 639 | |||
| GenBank mRNA | BC101908.1 | - | 5 | 639 | 7933832 | |
| CK476875.1 | - | 5 | 660 | |||
| FQ222101.1 | - | 4 | 482 | |||

Calu
Genomic Map