siRNA Details
| Gene Symbol: Calu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033898.1 | NP_001029070.1 | 7 | 915 | ||
| NM_022535.2 | NP_071980.2 | 7 | 915 | |||
| GenBank mRNA | AJ001929.1 | - | 7 | 948 | ||
| BC101908.1 | - | 7 | 915 | 7933832 | ||
| FQ222101.1 | - | 6 | 758 | |||

Calu
Genomic Map