siRNA Details
| Gene Symbol: Npw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153294.1 | NP_695206.1 | 1 | 500 | ||
| GenBank mRNA | AB084278.1 | - | 1 | 500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npw| Gene Symbol: Npw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153294.1 | NP_695206.1 | 1 | 500 | ||
| GenBank mRNA | AB084278.1 | - | 1 | 500 | ||