siRNA Details
| Gene Symbol: Lman2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106900.1 | NP_001100370.1 | 2 | 249 | ||
| GenBank mRNA | BC161950.1 | - | 233 | 9025901 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lman2l| Gene Symbol: Lman2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106900.1 | NP_001100370.1 | 2 | 249 | ||
| GenBank mRNA | BC161950.1 | - | 233 | 9025901 | ||