siRNA Details
| Gene Symbol: Pskh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108897.1 | NP_001102367.1 | 3 | 1188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pskh1| Gene Symbol: Pskh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108897.1 | NP_001102367.1 | 3 | 1188 | ||