siRNA Details
| Gene Symbol: Eapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106729.1 | NP_001100199.1 | 2 | 292 | ||
| NM_001134987.1 | NP_001128459.1 | 2 | 276 | |||
| XM_006240100.2 | XP_006240162.1 | 2 | 317 | |||
| GenBank mRNA | BC168880.1 | - | 2 | 276 | 9099474 | |

Eapp
Genomic Map