siRNA Details
| Gene Symbol: Fam195a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109475.1 | NP_001102945.1 | 4, 5 | 483 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam195a| Gene Symbol: Fam195a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109475.1 | NP_001102945.1 | 4, 5 | 483 | ||