siRNA Details
| Gene Symbol: LOC100361492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749104.3 | XP_003749152.1 | 7 | 1140 | ||
| Gene Symbol: Cyp2c24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271354.1 | NP_001258283.1 | 7 | 1140 | ||

LOC100361492
Genomic Map