siRNA Details
| Gene Symbol: PSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015310.3 | NP_056125.3 | 16 | 3046 | ||
| NM_206909.2 | NP_996792.1 | 13 | 1450 | |||
| XM_017013258.1 | XP_016868747.1 | 16 | 4355 | |||
| XM_017013259.1 | XP_016868748.1 | 16 | 4333 | |||
| XM_017013260.1 | XP_016868749.1 | 16 | 3106 | |||
| XM_017013261.1 | XP_016868750.1 | 15 | 3320 | |||
| XM_017013265.1 | XP_016868754.1 | 13 | 1856 | |||
| GenBank mRNA | AB023159.2 | - | 13 | 1431 | ||
| AF243495.2 | - | 14 | 1456 | |||
| AF519767.1 | - | 14 | 1404 | |||
| AK091317.1 | - | 16 | 3046 | |||
| AK296546.1 | - | 8 | 1064 | |||
| AK315099.1 | - | 13 | 1478 | |||
| AL832778.1 | - | 15 | 2913 | |||
| BC075044.2 | - | 13 | 1428 | 30915575 | ||
| BC075045.2 | - | 14 | 1431 | 30915359 | ||
| BX647406.1 | - | 8 | 1097 | |||
| GU727648.1 | - | 16 | 2946 | |||

PSD3
Genomic Map