siRNA Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234301.2 | XP_006234363.1 | 14, 15 | 5756 | ||
| XM_006234304.2 | XP_006234366.1 | 13, 14 | 5650 | |||
| XM_008761937.1 | XP_008760159.1 | 14, 15 | 4562 | |||
| GenBank mRNA | BC168937.1 | - | 2, 3 | 1029 | ||

Cobll1
Genomic Map