siRNA Details
| Gene Symbol: OBSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173431.1 | NP_001166902.1 | 13 | 4553 | ||
| NM_015311.2 | NP_056126.1 | 13 | 4553 | |||
| XM_005246424.4 | XP_005246481.1 | 12 | 4301 | |||
| XM_005246427.4 | XP_005246484.1 | 11 | 4016 | |||
| XM_011510857.2 | XP_011509159.1 | 13 | 4580 | |||
| XM_011510863.2 | XP_011509165.1 | 13 | 4572 | |||
| XM_017003696.1 | XP_016859185.1 | 13 | 4579 | |||
| XM_017003697.1 | XP_016859186.1 | 13 | 4579 | |||
| XM_017003698.1 | XP_016859187.1 | 12 | 4302 | |||
| XM_017003699.1 | XP_016859188.1 | 12 | 4303 | |||
| XM_017003700.1 | XP_016859189.1 | 11 | 4025 | |||
| GenBank mRNA | AK023854.1 | - | 4 | 944 | ||
| AK299261.1 | - | 4 | 845 | |||
| EF063637.1 | - | 13 | 4299 | |||
| EF063638.1 | - | 13 | 4296 | |||

OBSL1
Genomic Map