siRNA Details
| Gene Symbol: Tmem86b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109604.1 | NP_001103074.1 | 3 | 559 | ||
| XM_006228289.2 | XP_006228351.1 | 2 | 827 | |||
| GenBank mRNA | BC158795.1 | - | 2 | 1012 | 7314455 | |
| FQ219460.1 | - | 3 | 473 | |||

Tmem86b
Genomic Map