siRNA Details
| Gene Symbol: Ets2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107107.1 | NP_001100577.2 | 10 | 3418 | ||
| XM_006248177.2 | XP_006248239.1 | 10 | 3636 | |||
| GenBank mRNA | FQ214009.1 | - | 1 | 605 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ets2| Gene Symbol: Ets2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107107.1 | NP_001100577.2 | 10 | 3418 | ||
| XM_006248177.2 | XP_006248239.1 | 10 | 3636 | |||
| GenBank mRNA | FQ214009.1 | - | 1 | 605 | ||