siRNA Details
Gene Symbol: Gpkow | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109381.1 | NP_001102851.1 | 6 | 939 | ||
GenBank mRNA | BC166817.1 | - | 4 | 456 | 8365193 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpkow | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109381.1 | NP_001102851.1 | 6 | 939 | ||
GenBank mRNA | BC166817.1 | - | 4 | 456 | 8365193 |