siRNA Details
| Gene Symbol: Meig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134882.1 | NP_001128354.1 | 3 | 448 | ||
| XM_006254276.2 | XP_006254338.1 | 3 | 565 | |||
| XM_006254278.2 | XP_006254340.1 | 3 | 368 | |||
| XM_008771888.1 | XP_008770110.1 | 3 | 351 | |||

Meig1
Genomic Map