siRNA Details
| Gene Symbol: Tmem59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139465.2 | NP_001132937.1 | 8 | 1346 | ||
| GenBank mRNA | CV126271.1 | - | 2 | 523 | ||
| FM102590.1 | - | 1 | 350 | |||
| FQ212504.1 | - | 8 | 1312 | |||
| FQ231829.1 | - | 8 | 1332 | |||
| FQ232106.1 | - | 8 | 1351 | |||

Tmem59
Genomic Map