siRNA Details
| Gene Symbol: Olr840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000414.1 | NP_001000414.1 | 1 | 748 | ||
| Gene Symbol: LOC103692345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763692.1 | XP_008761914.1 | 1 | 586 | ||
| Gene Symbol: Olr838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000581.1 | NP_001000581.1 | 1 | 745 | ||

Olr840
Genomic Map