siRNA Details
| Gene Symbol: Fosl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013146.1 | NP_001013164.1 | 3 | 267 | ||
| NM_012954.1 | NP_037086.1 | 4 | 522 | |||
| GenBank mRNA | AY622611.1 | - | 4 | 519 | ||
| U18913.1 | - | 4 | 522 | |||
| U18982.1 | - | 3 | 267 | |||

Fosl2
Genomic Map