siRNA Details
| Gene Symbol: Ccdc176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134631.1 | NP_001128103.1 | 2 | 350 | ||
| XM_006240348.2 | XP_006240410.1 | 2 | 348 | |||
| XM_008764829.1 | XP_008763051.1 | 2 | 348 | |||
| XM_008764830.1 | XP_008763052.1 | 2 | 221 | |||

Ccdc176
Genomic Map