siRNA Details
| Gene Symbol: Zfp566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134726.1 | NP_001128198.1 | 1213 | |||
| GenBank mRNA | BC167072.1 | - | 1213 | 9028223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp566| Gene Symbol: Zfp566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134726.1 | NP_001128198.1 | 1213 | |||
| GenBank mRNA | BC167072.1 | - | 1213 | 9028223 | ||