siRNA Details
| Gene Symbol: Man2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134971.2 | NP_001128443.1 | 11 | 1774 | ||
| GenBank mRNA | BC168692.1 | - | 11 | 1775 | 7935016 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man2b2| Gene Symbol: Man2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134971.2 | NP_001128443.1 | 11 | 1774 | ||
| GenBank mRNA | BC168692.1 | - | 11 | 1775 | 7935016 | |