siRNA Details
| Gene Symbol: Slc7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134686.2 | NP_001128158.1 | 12 | 2290 | ||
| NM_022619.3 | NP_072141.2 | 12 | 2287 | |||
| GenBank mRNA | AF245001.1 | - | 12 | 2288 | ||
| AF245002.1 | - | 12 | 2291 | |||

Slc7a2
Genomic Map