siRNA Details
| Gene Symbol: Dhx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130039.1 | NP_001123511.1 | 12 | 2511 | ||
| XM_006230443.2 | XP_006230505.1 | 12 | 3076 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhx32| Gene Symbol: Dhx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130039.1 | NP_001123511.1 | 12 | 2511 | ||
| XM_006230443.2 | XP_006230505.1 | 12 | 3076 | |||