siRNA Details
| Gene Symbol: Myl12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135017.1 | NP_001128489.1 | 4 | 850 | ||
| XM_006245667.2 | XP_006245729.1 | 4 | 1004 | |||
| XM_006245668.2 | XP_006245730.1 | 4 | 945 | |||
| GenBank mRNA | FQ212603.1 | - | 2 | 554 | ||
| FQ215121.1 | - | 4 | 805 | |||
| FQ215776.1 | - | 995 | ||||
| FQ217399.1 | - | 4 | 896 | |||
| FQ217558.1 | - | 4 | 780 | |||
| FQ220614.1 | - | 836 | ||||
| FQ223132.1 | - | 4 | 814 | |||
| FQ228530.1 | - | 4 | 805 | |||
| FQ228551.1 | - | 1 | 275 | |||
| FQ229140.1 | - | 4 | 1059 | |||
| FQ232936.1 | - | 4 | 786 | |||
| X05566.1 | - | 4 | 790 | |||
| Z32519.1 | - | 4 | 788 | |||

Myl12a
Genomic Map