siRNA Details
| Gene Symbol: Gstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024304.1 | NP_001019475.1 | 1, 2 | 137 | ||
| GenBank mRNA | BC091199.1 | - | 1, 2 | 137 | 7379741 | |
| FJ179403.1 | - | 1, 2 | 41 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstm4| Gene Symbol: Gstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024304.1 | NP_001019475.1 | 1, 2 | 137 | ||
| GenBank mRNA | BC091199.1 | - | 1, 2 | 137 | 7379741 | |
| FJ179403.1 | - | 1, 2 | 41 | |||